missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexadecafluorosebacic Acid 96.0+%, TCI America™
$132.71 - $461.21
Chemical Identifiers
CAS | 307-78-8 |
---|---|
Molecular Formula | C10H2F16O4 |
Molecular Weight (g/mol) | 490.096 |
MDL Number | MFCD00042365 |
InChI Key | YPCSMEGZIYWAAZ-UHFFFAOYSA-N |
Synonym | Hexadecafluorodecanedioic Acid, Perfluorodecanedioic Acid, Perfluorosebacic Acid |
PubChem CID | 2733269 |
IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorodecanedioic acid |
SMILES | C(=O)(C(C(C(C(C(C(C(C(C(=O)O)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
Chemical Identifiers
307-78-8 | |
490.096 | |
YPCSMEGZIYWAAZ-UHFFFAOYSA-N | |
2733269 | |
C(=O)(C(C(C(C(C(C(C(C(C(=O)O)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
C10H2F16O4 | |
MFCD00042365 | |
Hexadecafluorodecanedioic Acid, Perfluorodecanedioic Acid, Perfluorosebacic Acid | |
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorodecanedioic acid |
Specifications
307-78-8 | |
White-Yellow | |
MFCD00042365 | |
Hexadecafluorodecanedioic Acid, Perfluorodecanedioic Acid, Perfluorosebacic Acid | |
C(=O)(C(C(C(C(C(C(C(C(C(=O)O)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O | |
490.096 | |
490.10 | |
Crystalline Powder |
162°C | |
C10H2F16O4 | |
5 g | |
YPCSMEGZIYWAAZ-UHFFFAOYSA-N | |
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorodecanedioic acid | |
2733269 | |
≥96.0% (GC,T) | |
Hexadecafluorosebacic Acid |
Safety and Handling
TSCA : No