missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexacosane, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 221220050

 View more versions of this product

Catalog No. AC221220050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Hexacosane
99%
Authentic
≥98.5%
C26H54
CH3(CH2)24CH3
MFCD00009354
0.8000g/mL
White
5g
630-01-3
Crystalline Solid
99%
Glass bottle
366.7
01, 175
56° to 59°C

Chemical Identifiers

630-01-3
C26H54
366.7
Hexacosane, 99%
99%
CH3(CH2)24CH3
MFCD00009354
≥98.5%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 211-124-1

TSCA : TSCA