Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexachloropropene, 95%

Chemical Identifiers

Chemical Name or Material Hexachloropropene
CAS 1888-71-7
Molecular Formula C3Cl6
Linear Formula Cl3CCCl=CCl2
Formula Weight 248.75
MDL Number MFCD00000835
UN Number UN3382
Beilstein 1766640
Synonym Perchloropropene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0780830

 
alfa aesar™
L0780830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0780818

 
alfa aesar™
L0780818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 248.75
Chemical Name or Material Hexachloropropene
Beilstein 1766640
TSCA TSCA
Health Hazard 2 GHS H Statement
H330-H315-H319-H335
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00000835
Molecular Formula C3Cl6
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P304+P340-P305+P351+P338-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 1888-71-7
UN Number UN3382
Synonym Perchloropropene
Melting Point −73°C
RTECS Number UD0175000
Boiling Point 209° to 210°C
Assay 95%
Linear Formula Cl3CCCl=CCl2
Quantity 250g
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, N.O.S.

Applications
Hexachloropropene is used as hydraulic fluid, solvent, plasticizer. It is also is used in organic synthesis.

Solubility
Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P304+P340-P305+P351+P338-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330-H315-H319-H335
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.