Please login to your online account to display your discounted pricing

Hexachloroplatinic(IV) Acid Solution (about 10%), (3.8% Pt) GR For analysis, EMD Millipore™


Manufacturer: emd millipore corporation  1073410005

 View more versions of this product

Catalog No. M1073410005

  • / Each
 

Description & Specifications

Chemical Identifiers

DOT Information DOT Class: 8, Packaging Group: III
Chemical Name or Material Hexachloroplatinic(IV) acid solution about
Odor Odorless
Packaging Glass bottle
Physical Form Liquid
Assay Percent Range 10%
Appearance Orange liquid
CAS 16941-12-1
UN Number UN 3264
Health Hazard 1 H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.
H334: May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Specifications

Chemical Name or Material Hexachloroplatinic(IV) acid solution about
CAS 16941-12-1
UN Number UN 3264
Accessories

H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.
H334: May cause allergy or asthma symptoms or breathing difficulties if inhaled.