Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexachloro-1,3-butadiene, 97%

Chemical Identifiers

Chemical Name or Material Hexachloro-1,3-butadiene
CAS 87-68-3
Molecular Formula CCl2=CClCCl=CCl2
Formula Weight 260.76
MDL Number MFCD00000836
UN Number UN2279
Beilstein 1766570
Merck Index 14,4678
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA173160B

  SDS
alfa aesar™
A173160B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1731618

  SDS
alfa aesar™
A1731618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1731630

  SDS
alfa aesar™
A1731630
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 260.76
Odor Mild
Beilstein 1766570
Chemical Name or Material Hexachloro-1,3-butadiene
TSCA TSCA
Assay 97%
Molecular Formula CCl2=CClCCl=CCl2
MDL Number MFCD00000836
CAS 87-68-3
UN Number UN2279
Merck Index 14,4678
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H310-H330-H351-H361-H315-H319
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: HEXACHLOROBUTADIENE
RTECS Number EJ0700000
Boiling Point 215°C
Melting Point −21°C
Quantity 1000g
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H310-H330-H351-H361-H315-H319
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.