Please login to your online account to display your discounted pricing

Hexacarbonyltungsten, 97%, Alfa Aesar™

$102.68 - $102.68

Chemical Identifiers

CAS 14040-11-0
Physical Form Crystalline
Assay Percent Range 97%
Molecular Formula C6O6W
Formula Weight 351.91
MDL Number MFCD00011462
Synonym Tungsten hexacarbonyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4320518

 
alfa aesar
4320518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4320509

 
alfa aesar
4320509
10g Each for $102.68
Description & Specifications

Specifications

Formula Weight 351.91
Quantity 50g
Color White
Physical Form Crystalline
Assay Percent Range 97%
Odor Nearly odorless
Molecular Formula C6O6W
MDL Number MFCD00011462
CAS 14040-11-0
Synonym Tungsten hexacarbonyl
Vapor Pressure 1.596hPa (1mm Hg) at 67°C