missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexacarbonylmolybdenum, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1305722

 View more versions of this product

Catalog No. AA1305722



Specifications

Specifications

Hexacarbonylmolybdenum
White
100g
13939-06-5
C6MoO6
MFCD00003466
molybdenumhexacarbonyl, carbon monooxide-molybdenum 6:1, hexakis carbon monoxide molybdenum, hexacarbonylmolybdenu, hexakis methanidylidyneoxidanium molybdenum, hexacarbonylmolybdenum, may contain up to cr 5g
KMKBZNSIJQWHJA-UHFFFAOYSA-N
carbon monoxide;molybdenum
2724068
264
98%
1.96g/mL
~150°C (decomposition)
Up to 1% Cr
98%
Mo(CO)6
UN3466
May contain up to 1% Cr
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mo]
264.01
CHEBI:30508
Crystalline
May contain up to 1% Cr

Chemical Identifiers

13939-06-5
264.01
KMKBZNSIJQWHJA-UHFFFAOYSA-N
2724068
carbon monoxide;molybdenum
C6MoO6
MFCD00003466
molybdenumhexacarbonyl, carbon monooxide-molybdenum 6:1, hexakis carbon monoxide molybdenum, hexacarbonylmolybdenu, hexakis methanidylidyneoxidanium molybdenum, hexacarbonylmolybdenum, may contain up to cr 5g
CHEBI:30508
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mo]
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.

GHS P Statement
P280f-P304+P340-P310-P302+P352
Wear protective clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.

TSCA : TSCA

SDS Documents