Learn More
Hexaammineruthenium(III) chloride, Ru 32.1% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01051106
Description
Depending on the choice of suitable reductive or oxidative quencher this catalyst can be used to trigger photoreduction or photooxidation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexaammineruthenium(III) chloride | |
Yellow | |
MFCD00011478 | |
UN3288 | |
Very soluble in water. | |
N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Ru+3] | |
309.61 | |
309.61 | |
Powder |
14282-91-8 | |
Cl3H18N6Ru | |
5 g | |
hexaammineruthenium iii chloride, hexaammineruthenium trichloride, hexaamminetrichlororuthenium, hexaammineruthenium trichloride, hydrate, ruthenium 3+ , hexaammine-, trichloride, hydrate, ruthenium 3+ chloride ammoniate 1:3:6, hexaammineruthenium iii trichloride, ruthenium 3+ ion hexaamine trichloride, ruthenium 3+ , hexaammine-, trichloride | |
GBDZMMXUOBAJMN-UHFFFAOYSA-K | |
azane;ruthenium(3+);trichloride | |
159731 | |
Ru 32.1% min. |
Chemical Identifiers
14282-91-8 | |
309.61 | |
GBDZMMXUOBAJMN-UHFFFAOYSA-K | |
159731 |
Cl3H18N6Ru | |
MFCD00011478 | |
hexaammineruthenium iii chloride, hexaammineruthenium trichloride, hexaamminetrichlororuthenium, hexaammineruthenium trichloride, hydrate, ruthenium 3+ , hexaammine-, trichloride, hydrate, ruthenium 3+ chloride ammoniate 1:3:6, hexaammineruthenium iii trichloride, ruthenium 3+ ion hexaamine trichloride, ruthenium 3+ , hexaammine-, trichloride | |
N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Ru+3] |
Safety and Handling
GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber : 238-176-8
RTECSNumber : VM2651530
TSCA : No
storageNote1 : 2°C to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only