missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexaamminenickel(II) iodide, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4448122

 View more versions of this product

Catalog No. AA4448122



Specifications

Specifications

Hexaamminenickel (II) iodide
13859-68-2
Ni(NH3)6I2
diiodonickel-ammonia 1/6
Crystalline
100g
(metals basis)
MFCD00058808
414.68
99.999%

Chemical Identifiers

13859-68-2
Ni(NH3)6I2
MFCD00058808
diiodonickel-ammonia 1/6
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H350

TSCA : No

Recommended Storage : Ambient temperatures

Documents