Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexaamminecobalt(III) nitrate

Chemical Identifiers

Chemical Name or Material Hexaamminecobalt(III) nitrate
CAS 10534-86-8
Linear Formula [Co(NH3)6](NO3)3
Formula Weight 347.13
MDL Number MFCD00061421
UN Number UN1477
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2255506

  SDS
alfa aesar™
2255506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2255514

  SDS
alfa aesar™
2255514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 347.13
Chemical Name or Material Hexaamminecobalt(III) nitrate
TSCA TSCA
Physical Form Powder
Linear Formula [Co(NH3)6](NO3)3
MDL Number MFCD00061421
CAS 10534-86-8
UN Number UN1477
Health Hazard 1 Danger
Density 1.804g/mL
Health Hazard 2 GHS H Statement
H272-H351-H317
May intensify fire
oxidizer.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P221-P210-P220-P363-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P363-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H351-H317
May intensify fire
oxidizer.
Suspected of causing cancer.
May cause an allergic skin reaction.