missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexaamminecobalt(III) chloride, 99%

Manufacturer:  Alfa Aesar™ A1547036

 View more versions of this product

Catalog No. AAA1547036



Description

Description

Applications
Hexaamminecobalt(III) chloride is a component of structural biology methods, especially for DNA or RNA, where the positive ions stabilize the tertiary structure of the phosphate backbone, which favors to solve their structure by X-ray crystallography. It serves as an excellent antibacterial and antiviral property.

Solubility
Soluble in ammonia and water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Hexaamminecobalt(III) chloride
500g
H18Cl3CoN6
MFCD00036304
267.48
Odorless
10534-89-1
[Co(NH3)6]Cl3
14,4675
99%

Chemical Identifiers

10534-89-1
H18Cl3CoN6
MFCD00036304
Safety and Handling

Safety and Handling

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents