missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexaamminecobalt(III) chloride, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1376914

 View more versions of this product

Catalog No. AA1376914



Specifications

Specifications

Hexaamminecobalt(III) chloride
97%
MFCD00036304
267.48
1.71g/cm3
Orange
10534-89-1
[Co(NH3)6]Cl3
Orange powder
Crystalline
Odorless
25g

Chemical Identifiers

10534-89-1
[Co(NH3)6]Cl3
MFCD00036304
Safety and Handling

Safety and Handling

H351-H317; Suspected of causing cancer. May cause an allergic skin reaction.

EINECSNumber : 234-103-9

SDS Documents