Learn More
Hexa-n-butylditin, 98%, ACROS Organics™
Supplier: Thermo Scientific Chemicals 351510500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexa-n-butylditin | |
Colorless to Yellow | |
197.0°C to 198.0°C (10.0 mmHg) | |
Authentic | |
Glass Bottle | |
1.5100 to 1.5130 | |
MFCD00009417 | |
1.148 | |
REDSKZBUUUQMSK-UHFFFAOYSA-N | |
tributyltin | |
6327815 | |
97% |
813-19-4 | |
1.1480g/mL | |
>110°C | |
97% | |
C24H54Sn2 | |
([CH3(CH2)3]3Sn-)2 | |
50 g | |
hexabutylditin, hexabutyldistannane, bis, distannane, hexabutyl, tin, hexabutyldi, stannane, hexabutyldi, hexa-n-butylditin, 1,1,1,2,2,2-hexabutyldistannane, hexabutyidistannane, hexabutyl distannane | |
CCCC[Sn](CCCC)(CCCC)[Sn](CCCC)(CCCC)CCCC | |
580.12 | |
580.08 | |
Liquid |
Chemical Identifiers
813-19-4 | |
580.12 | |
REDSKZBUUUQMSK-UHFFFAOYSA-N | |
6327815 | |
CCCC[Sn](CCCC)(CCCC)[Sn](CCCC)(CCCC)CCCC |
C24H54Sn2 | |
MFCD00009417 | |
hexabutylditin, hexabutyldistannane, bis, distannane, hexabutyl, tin, hexabutyldi, stannane, hexabutyldi, hexa-n-butylditin, 1,1,1,2,2,2-hexabutyldistannane, hexabutyidistannane, hexabutyl distannane | |
tributyltin |
Safety and Handling
GHS H Statement:
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 212-383-3
RUO – Research Use Only