missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexa-n-butylditin, 98%

$55.11 - $55.11

Chemical Identifiers

CAS 813-19-4
Molecular Formula C24H54Sn2
Linear Formula (CH3CH2CH2CH2)3Sn-Sn(CH2CH2CH2CH3)3
Formula Weight 580.07
MDL Number MFCD00009417
UN Number UN2788
Beilstein 3605476
Synonym Bis(tri-n-butyltin); Hexa-n-butyldistannane
Chemical Name or Material Hexa-n-butylditin
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1200709 SDS Alfa Aesar™
A1200709*
10g Each for $55.11
AAA1200718 SDS Alfa Aesar™
A1200718
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1200730 SDS Alfa Aesar™
A1200730
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

813-19-4
(CH3CH2CH2CH2)3Sn-Sn(CH2CH2CH2CH3)3
MFCD00009417
3605476
Hexa-n-butylditin
C24H54Sn2
580.07
UN2788
Bis(tri-n-butyltin); Hexa-n-butyldistannane

Specifications

197° to 198°C (10mmHg)
10g
C24H54Sn2
580.07
UN2788
Bis(tri-n-butyltin); Hexa-n-butyldistannane
98%
813-19-4
124°C (255°F)
(CH3CH2CH2CH2)3Sn-Sn(CH2CH2CH2CH3)3
MFCD00009417
3605476
Hexa-n-butylditin

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

Recommended Storage : −30°C to −10°C