Heterogeneous Catalyst Kit, 40 sample
Specifications
Synonym Global Heterogeneous Catalyst Kit, 40 sample
Product Type Catalyst
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH37163KT Alfa Aesar™
H37163KT
1 Kit N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
Excellent tools for screening and research projects. Provides instant availability to 40 catalyst types with varying characteristics and metal distribution. The kit is supplied with an Applications Table that allows for easy identification of prime candidates for catalytic processes.
Description & Specifications
Specifications
Global Heterogeneous Catalyst Kit, 40 sample
Catalyst