missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hesperidin, 97%, includes analogeous compounds, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 123460050
Specifications
Hesperidin | |
Authentic | |
Glass Bottle | |
Brown to White | |
5g | |
520-26-3 | |
C28H34O15 | |
15, 4706 | |
QUQPHWDTPGMPEX-DPTDPGGASA-N | |
(2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxy-2,3-dihydrochromen-4-one | |
45358131 | |
Powder |
20ppm max | |
4% max. | |
− 76.00 | |
258.0°C to 262.0°C | |
97% | |
97% | |
MFCD00075663 | |
Solubility in water: 1g/50l. Other solubilities: soluble in diluted alkalies,soluble in pyridine,soluble in formamide and dimethylformamide (60 c),slightly sol. in methanol and glacial acetic acid,practically insol. in acetone benzene and chcl3 | |
CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=O)CC(OC4=C3)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O | |
610.54 | |
610.54 | |
97% |
Chemical Identifiers
520-26-3 | |
610.54 | |
QUQPHWDTPGMPEX-DPTDPGGASA-N | |
(2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxy-2,3-dihydrochromen-4-one |
C28H34O15 | |
MFCD00075663 | |
45358131 | |
CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=O)CC(OC4=C3)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 208-288-1
RTECSNumber : MK6650000
TSCA : TSCA