missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hesperidin, 95%

Acts as an antioxidant

$61.69 - $185.05

Chemical Identifiers

CAS 520-26-3
Molecular Formula C28H34O15
Molecular Weight (g/mol) 610.565
MDL Number MFCD00075663
InChI Key QUQPHWDTPGMPEX-DPTDPGGASA-N
Synonym Hesperetin 7-rhamnoglucoside; Hesperitin-7-rutinoside
PubChem CID 45358131
IUPAC Name (2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxy-2,3-dihydrochromen-4-one
SMILES CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=O)CC(OC4=C3)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6212614 View Documents Alfa Aesar™
J6212614
25g Each for $61.69
 
AAJ6212622 View Documents Alfa Aesar™
J6212622
100g Each for $185.05
 
Description

Description

Applications
Hesperidin which has been found to be a potent chemopreventive agent in oral, urinary bladder, and colon carcinogenesis in animal models. Applied in food field, it is used as a flavour enhancer and sweetener in a wide variety of alcoholicbeverage, dessert foods and savory foods, applied in cosmetics, it can be added into toothpaste and mouth wash, applied in pharmaceutical field, it is mainly used to reducing the bitterness of in form. It also enhance the action of Vitamin C.

Solubility
Insoluble in water. Soluble in organic solvents such as DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents. Keep away from strong light and heat.
Specifications

Chemical Identifiers

520-26-3
610.565
QUQPHWDTPGMPEX-DPTDPGGASA-N
45358131
CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=O)CC(OC4=C3)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O
C28H34O15
MFCD00075663
Hesperetin 7-rhamnoglucoside; Hesperitin-7-rutinoside
(2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxy-2,3-dihydrochromen-4-one

Specifications

Hygroscopic
250°C to 255°C
520-26-3
MFCD00075663
14,4671
QUQPHWDTPGMPEX-DPTDPGGASA-N
(2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxy-2,3-dihydrochromen-4-one
45358131
Powder
Hesperidin
Brown to White
25g
C28H34O15
75140
Hesperetin 7-rhamnoglucoside; Hesperitin-7-rutinoside
CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=O)CC(OC4=C3)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O
610.565
610.56
95%
Safety and Handling

Safety and Handling

RTECSNumber : MK6650000

TSCA : TSCA

Recommended Storage : 2° to 8°C