missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hesperidin, 95%, Thermo Scientific Chemicals

Acts as an antioxidant

Supplier:  Thermo Scientific Chemicals J6212622

 View more versions of this product

Catalog No. AAJ6212622



Description

Description

Hesperidin which has been found to be a potent chemopreventive agent in oral, urinary bladder, and colon carcinogenesis in animal models. Applied in food field, it is used as a flavour enhancer and sweetener in a wide variety of alcoholicbeverage, dessert foods and savory foods, applied in cosmetics, it can be added into toothpaste and mouth wash, applied in pharmaceutical field, it is mainly used to reducing the bitterness of in form. It also enhance the action of Vitamin C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hesperidin which has been found to be a potent chemopreventive agent in oral, urinary bladder, and colon carcinogenesis in animal models. Applied in food field, it is used as a flavour enhancer and sweetener in a wide variety of alcoholicbeverage, dessert foods and savory foods, applied in cosmetics, it can be added into toothpaste and mouth wash, applied in pharmaceutical field, it is mainly used to reducing the bitterness of in form. It also enhance the action of Vitamin C.

Solubility
Insoluble in water. Soluble in organic solvents such as DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents. Keep away from strong light and heat.
Specifications

Specifications

Hesperidin
250°C to 255°C (decomposition)
C28H34O15
100 g
Hygroscopic
Hesperetin 7-rhamnoglucoside; Hesperitin-7-rutinoside
QUQPHWDTPGMPEX-QJBIFVCTSA-N
(2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-3,4-dihydro-2H-1-benzopyran-4-one
45358131
95%
520-26-3
Brown to White
MFCD00075663
75140
14,4671
Insoluble in water. Soluble in organic solvents such as DMSO.
COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1
610.57
610.56
Powder

Chemical Identifiers

520-26-3
610.57
QUQPHWDTPGMPEX-QJBIFVCTSA-N
45358131
COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1
C28H34O15
MFCD00075663
Hesperetin 7-rhamnoglucoside; Hesperitin-7-rutinoside
(2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-3,4-dihydro-2H-1-benzopyran-4-one
Safety and Handling

Safety and Handling

EINECSNumber : 208-288-1

RTECSNumber : MK6650000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only