Please login to your online account to display your discounted pricing

Hesperadin hydrochloride, Tocris™

Potent Aurora kinase B inhibitor

$335.85 - $1,384.48

Chemical Identifiers

Chemical Name or Material Hesperadin hydrochloride
Assay Percent Range >99%
Molecular Formula C29H32N4O3S·HCl
Formula Weight 553.12
Synonym N-[2, 3-Dihydro-2-oxo-3-[(3Z)-phenyl[[4-(1-piperidinylmethyl)phenyl]amino]methylene]-1H-indol-5-yl]-ethanesulfonamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

398850

 
r & d systems
RDS398850
50mg Each for $1,384.48

398810

 
r & d systems
RDS398810
10mg Each for $335.85
Description & Specifications

Specifications

Formula Weight 553.12
Quantity 50mg
Chemical Name or Material Hesperadin hydrochloride
Assay Percent Range >99%
Molecular Formula C29H32N4O3S·HCl
CAS 422513-13-1
Synonym N-[2, 3-Dihydro-2-oxo-3-[(3Z)-phenyl[[4-(1-piperidinylmethyl)phenyl]amino]methylene]-1H-indol-5-yl]-ethanesulfonamide hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature