Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Heregulin/NDF/GGF/Neuregulin Ab-2, Rabbit Polyclonal Antibody

Recommended for Western Blotting and Biological Blockade, Thermo Scientific™ Heregulin/NDF/GGF/Neuregulin Ab-2, Rabbit Polyclonal Antibody provides accurate, reproducible results. Rabbit Polyclonal Antibody.

$277.12 - $1,619.02

Specifications

Antigen Heregulin/NDF/GGF/Neuregulin Ab-2
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB276B

 
thermo scientific™ lab vision™
RB276B
Biotinylated Ab with BSA and Azide 1mL Each for $1,619.02

RB276B0

 
thermo scientific™ lab vision™
RB276B0
Biotinylated Ab with BSA and Azide 100µL Each for $370.65

RB276B1

 
thermo scientific™ lab vision™
RB276B1
Biotinylated Ab with BSA and Azide 500µL Each for $1,010.08

RB276P

 
thermo scientific™ lab vision™
RB276P
Purified Ab with BSA and Azide 1mL Each for $1,297.63

RB276P0

 
thermo scientific™ lab vision™
RB276P0
Purified Ab with BSA and Azide 100µL Each for $277.12

RB276P1

 
thermo scientific™ lab vision™
RB276P1
Purified Ab with BSA and Azide 500µL Each for $776.88

RB276P1ABX

 
thermo scientific™ lab vision™
RB276P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $854.04

RB276PABX

 
thermo scientific™ lab vision™
RB276PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,403.82
Description & Specifications

Specifications

Antigen Heregulin/NDF/GGF/Neuregulin Ab-2
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline EGFR Family
Type Primary
Immunogen A highly antigenic synthetic peptide from the EGF-like domain of human heregulin
Isotype IgG

Recommended for Western Blotting and Biological Blockade, Thermo Scientific™ Heregulin/NDF/GGF/Neuregulin Ab-2, Rabbit Polyclonal Antibody provides accurate, reproducible results. Rabbit Polyclonal Antibody.

Heregulin (HRG) or neu differentiation factor (NDF) binds to c-erbB-3 and c-erbB-4 receptors on mammary and neuronal cells with low and high affinities, respectively. HRG/NDF exists in several isoforms, which are classified in two groups, α and β that differ in their EGF-like domain and in receptor binding affinity.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not known.

Epitope: EGF-like domain

Immunogen: A highly antigenic synthetic peptide from the EGF-like domain of human heregulin

Molecular Weight: 44kDa

Positive Control: HRGβ1 recombinant protein or LS174T cells

Cellular Localization: Cytoplasmic and intercellular spaces

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Biological Blockade

Note:

USA: RUO; Int'l: RUO