Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Heregulin/NDF/GGF/Neuregulin Ab-2, Rabbit Polyclonal Antibody


Recommended for Western Blotting and Biological Blockade, Thermo Scientific™ Heregulin/NDF/GGF/Neuregulin Ab-2, Rabbit Polyclonal Antibody provides accurate, reproducible results. Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB276P1ABX

 View more versions of this product

Catalog No. RB276P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG
Format Concentrated
Immunogen A highly antigenic synthetic peptide from the EGF-like domain of human heregulin
Antigen Heregulin/NDF/GGF/Neuregulin Ab-2
Host Species Rabbit
Regulatory Status RUO
Quantity 500µL
Applications Western Blotting
Formulation Ab without BSA/Azide (Frozen)
Classification Polyclonal
Conjugate Unlabeled
Research Discipline EGFR Family
Concentration 1mg/mL
Type Primary

Recommended for Western Blotting and Biological Blockade, Thermo Scientific™ Heregulin/NDF/GGF/Neuregulin Ab-2, Rabbit Polyclonal Antibody provides accurate, reproducible results. Rabbit Polyclonal Antibody.

Heregulin (HRG) or neu differentiation factor (NDF) binds to c-erbB-3 and c-erbB-4 receptors on mammary and neuronal cells with low and high affinities, respectively. HRG/NDF exists in several isoforms, which are classified in two groups, α and β that differ in their EGF-like domain and in receptor binding affinity.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not known.

Epitope: EGF-like domain

Immunogen: A highly antigenic synthetic peptide from the EGF-like domain of human heregulin

Molecular Weight: 44kDa

Positive Control: HRGβ1 recombinant protein or LS174T cells

Cellular Localization: Cytoplasmic and intercellular spaces

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Biological Blockade

Note:

USA: RUO; Int'l: RUO