Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Heregulin/NDF/GGF/Neuregulin Ab-1, Mouse Monoclonal Antibody

Recommended for Western Blotting and Immunoprecipitation, Thermo Scientific™ Heregulin/NDF/GGF/Neuregulin Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

$272.02 - $1,373.31

Specifications

Antigen Heregulin/NDF/GGF/Neuregulin Ab-1
Clone 7D5
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS272P

 
thermo scientific™ lab vision™
MS272P
Purified Ab with BSA and Azide 1mL Each for $1,263.76

MS272P0

 
thermo scientific™ lab vision™
MS272P0
Purified Ab with BSA and Azide 100µL Each for $272.02

MS272P1

 
thermo scientific™ lab vision™
MS272P1
Purified Ab with BSA and Azide 500µL Each for $762.59

MS272P1ABX

 
thermo scientific™ lab vision™
MS272P1ABX
Ab Without BSA/Azide (Frozen) 100µL Each for $832.80

MS272PABX

 
thermo scientific™ lab vision™
MS272PABX
Ab Without BSA/Azide (Frozen) 200µL Each for $1,373.31

MS272R7

 
thermo scientific™ lab vision™
MS272R7
Ready-to-use for IHC (UVLP) 7mL Each for $751.12
Description & Specifications

Specifications

Antigen Heregulin/NDF/GGF/Neuregulin Ab-1
Clone 7D5
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Mouse
Regulatory Status RUO
Research Discipline EGFR Family
Type Primary
Immunogen Recombinant extracellular domain of rat NDF protein
Isotype IgG2a

Recommended for Western Blotting and Immunoprecipitation, Thermo Scientific™ Heregulin/NDF/GGF/Neuregulin Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Heregulin (HRG) or neu differentiation factor (NDF) binds to c-erbB-3 and c-erbB-4 receptors on mammary and neuronal cells with low and high affinities, respectively. HRG/NDF exists in several isoforms, which are classified in two groups, α and β that differ in their EGF-like domain and in receptor binding affinity.

Host Species: Mouse

Clone: 7D5

Isotype: IgG2a

Species Reactivity: Human, Rat and Mouse.

Epitope: Extracellular

Immunogen: Recombinant extracellular domain of rat NDF protein

Molecular Weight: 44kDa

Positive Control: HRGα1 or HRGβ1 recombinant protein, Prostate CA

Cellular Localization: Cytoplasmic and intercellular spaces

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO