missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Heregulin/NDF/GGF/Neuregulin Ab-1, Mouse Monoclonal Antibody

Recommended for Western Blotting and Immunoprecipitation, Thermo Scientific™ Heregulin/NDF/GGF/Neuregulin Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS272P1ABX

 View more versions of this product

Catalog No. MS272P1ABX


 

Specifications

Description

Recommended for Western Blotting and Immunoprecipitation, Thermo Scientific™ Heregulin/NDF/GGF/Neuregulin Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Heregulin (HRG) or neu differentiation factor (NDF) binds to c-erbB-3 and c-erbB-4 receptors on mammary and neuronal cells with low and high affinities, respectively. HRG/NDF exists in several isoforms, which are classified in two groups, α and β that differ in their EGF-like domain and in receptor binding affinity.

Host Species: Mouse

Clone: 7D5

Isotype: IgG2a

Species Reactivity: Human, Rat and Mouse.

Epitope: Extracellular

Immunogen: Recombinant extracellular domain of rat NDF protein

Molecular Weight: 44kDa

Positive Control: HRGα1 or HRGβ1 recombinant protein, Prostate CA

Cellular Localization: Cytoplasmic and intercellular spaces

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Heregulin/NDF/GGF/Neuregulin Ab-1
Monoclonal
1mg/mL
Ab Without BSA/Azide (Frozen)
Murine
IgG2a
RUO
Primary
Human,Murine,Rat
Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting
7D5
Unlabeled
Concentrated
Recombinant extracellular domain of rat NDF protein
100μL
EGFR Family
Monoclonal