Please login to your online account to display your discounted pricing

Alfa Aesar™ Herbimycin A, 95%

Chemical Identifiers

Chemical Name or Material Herbimycin A
CAS 70563-58-5
Formula Weight 574.66
MDL Number MFCD00151702
Beilstein 4834067
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66427MCR

  SDS
alfa aesar™
J66427MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 574.66
CAS 70563-58-5
Chemical Name or Material Herbimycin A
MDL Number MFCD00151702
Beilstein 4834067
Physical Form Powder
Assay 95%
Solubility Information Soluble in DMSO
Boiling Point 752°C
Color Yellow
Storage Note 1 -30° to -10°C
Quantity 1mg
Sensitivity Light Sensitive & Hygroscopic
RTECS Number LX8930000

  • A sparingly soluble benzoquinone anti-tumor agent which interacts with the stress induced chaperone protein Hsp90 (heat shock protein 90)

Safety and Handling

-30° to -10°C