Thermo Scientific™ Heracell™ 150i and 240i Tri-Gas Incubators
Protect valuable samples while optimizing cell growth. These incubators offer convenient oxygen control, in noncorrosive stainless steel or 100% pure copper.
Specifications
Cabinet Material cold rolled steel
CO2 Concentration Range 0 to 20%
Data Outputs RS232
Humidity Delivery integral panless system
O2 Sensor Technology zirconium oxide
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) CO2 Sensor Technology Chamber Material Oxygen Control Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) CO2 Sensor Technology Chamber Material Oxygen Control Price Quantity  
13-998-039 Accessories Thermo Scientific™
51026402
150L IR Electropolished stainless steel 1-21% N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-998-038 Thermo Scientific™
51026408
150L TCD Solid copper 1-21% N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-998-037 Thermo Scientific™
51026410
150L TCD Electropolished stainless steel 1-21% N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-998-048 Thermo Scientific™
51026422
240L TC Solid copper 1-21% N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-998-047 Thermo Scientific™
51026423
240L TC Electropolished stainless steel 1-21% N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-998-041 Thermo Scientific™
51026529
150L TCD Electropolished stainless steel 5-90% N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-998-050 Thermo Scientific™
51026533
240L IR Solid copper 1-21% N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-998-042 Thermo Scientific™
51026536
150L TCD Solid copper 5-90% N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-998-040 Thermo Scientific™
51026537
150L IR Solid copper 1-21% N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-998-049 Accessories Thermo Scientific™
51026556
240L IR Electropolished Stainless-steel 1-21% N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
Protect valuable samples while optimizing cell growth with Thermo Scientific™ Heracell™ 150i and 240i Tri-Gas Incubators. They allow convenient oxygen control and are available in noncorrosive stainless steel or 100% pure copper. The incubators provide the ideal in vitro environment and are clean, reliable and easy to use.
iCAN™ Touchscreen Control
  • Door-mounted for easy accessibility and viewing
  • Convenient on-screen user prompts for simplified measurement and control navigation
  • Choice of languages
  • Changes in culture environment visible on screen logs and usage recording
  • Visual monitor alarm
TC or IR CO2 Sensor
  • Standard thermal conductivity (TC) sensor features accurate gas sensor technology
  • Infrared (IR) sensor features exclusive split-beam technology for precise CO2 control in fluctuating temperature and humidity conditions
Rapid Humidity Recovery
  • Large, directly heated water reservoir
  • Recovery times are up to five times faster than conventional water pans
  • Water level alarm with audible and visual alerts
Optional O2 Control with Auto-Cal
  • Two O2 control ranges precisely control oxygen concentration
  • Ideal for applications requiring decreased or increased O2 conditions
  • Zirconium Oxide sensor
Description & Specifications
Specifications
cold rolled steel
RS232
zirconium oxide
0 to 20%
integral panless system