Please login to your online account to display your discounted pricing
Thermo Scientific™ Heracell™ 150i and 240i CO2 Incubators with Stainless-Steel Chambers
Protect valuable samples while optimizing cell growth. These CO2 incubators with non-corrosive stainless-steel interior chambers provide the ideal in-vitro environment.
Specifications
Chamber Material Electropolished Stainless-steel
CO2 Concentration Range 0 to 20%
Data Outputs RS232
Disinfection Time 90°C/9 hr.
Humidity Delivery integral panless system
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) CO2 Sensor Technology Voltage Price Quantity    
13-998-051
thermo scientific™
50116048
0.07 x 5.3 cu. ft. TC 120V CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-998-033
thermo scientific™
51026282
150L TC 120V CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-998-043
thermo scientific™
51026331
240L TC 120V CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-998-035
  Accessories
thermo scientific™
51026406
150L IR 120V CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-998-045
  Accessories
thermo scientific™
51026420
240L IR 120V CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Chamber Material Electropolished Stainless-steel
CO2 Concentration Range 0 to 20%
Data Outputs RS232
Disinfection Time 90°C/9 hr.
Humidity Delivery integral panless system
Oxygen Control Optional 1-21% or 5-90%
Relative Humidity to 95%
Temperature Range (Metric) Ambient +3° to 55°C
Hertz 50/60Hz
Shelf Material Electropolished Stainless-steel
Protect valuable samples while optimizing cell growth. Thermo Scientific™ Heracell™ 150i and 240i CO2 Incubators with Stainless-Steel Chambers provide the ideal in-vitro environment. Clean, reliable and easy to use, they feature fast recovery characteristics and a convenient touchscreen user-interface.
iCAN™ Touchscreen Control
  • Door-mounted for easy accessibility and viewing
  • Convenient on-screen user prompts for simplified measurement and control navigation
  • Choice of languages
  • Changes in culture environment visible on screen logs and usage recording
  • Visual monitor alarm
TC or IR CO2 Sensor
  • Standard thermal conductivity (TC) sensor features accurate gas sensor technology
  • Infrared (IR) sensor features exclusive split-beam technology for precise CO2 control in fluctuating temperature and humidity conditions
Rapid Humidity Recovery
  • Large, directly heated water reservoir
  • Recovery times are up to five times faster than conventional water pans
  • Water level alarm with audible and visual alerts
O2 Control with Auto-Cal
  • Two O2 control ranges precisely control oxygen concentration
  • Ideal for applications requiring decreased or increased O2 conditions