missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-4/c-erbB-4, Rabbit Polyclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin) and Western Blotting, Thermo Scientific™ HER-4/c-erbB-4, Rabbit Polyclonal Antibody provides accurate, reproducible results.

$277.92 - $1,351.39

Specifications

Antigen HER-4/c-erbB-4
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
RB9045P Thermo Scientific™ Lab Vision™
RB9045P
Purified Ab with BSA and Azide 1mL Each for $1,351.39
RB9045P0 Thermo Scientific™ Lab Vision™
RB9045P0
Purified Ab with BSA and Azide 100μL Each for $277.92
RB9045P1 Thermo Scientific™ Lab Vision™
RB9045P1
Purified Ab with BSA and Azide 500μL Each for $761.61
RB9045R7 Thermo Scientific™ Lab Vision™
RB9045R7
Ready-to-use for IHC (UVLP) 7mL Each for $745.94
Specifications

Description

Recommended for Immunohistochemistry (Formalin/paraffin) and Western Blotting, Thermo Scientific™ HER-4/c-erbB-4, Rabbit Polyclonal Antibody provides accurate, reproducible results.

c-erbB-4 is the fourth member of class I receptor kinase family. HER-4 is most predominantly expressed in several breast carcinoma cell lines, and in normal skeletal muscle, heart, pituitary, brain, and cerebellum. Breast tumor cell lines T47-D, MDA-MB-453, BT-474 and H3396 are found to have the highest levels of mRNA, and intermediate levels are seen in MCF-7, MDA-MB-330 and MDA-MB-361. Expression of erbB-4 is low or absent in some breast tumor cell lines such as MDA-MB-231, MDA-MB-157, MDA-MB-468, and SKBR-3.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human HER-4 protein

Molecular Weight: 185kDa

Positive Control: Breast CA T47D cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

HER-4/c-erbB-4
Polyclonal
Human
RUO
Primary
IgG
Western Blot
Unlabeled
Rabbit
EGFR Family
A synthetic peptide from C-terminus of human c-erbB-4 protein