Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-4/c-erbB-4, Mouse Monoclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ HER-4/c-erbB-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

$271.02 - $1,283.88

Specifications

Antigen HER-4/c-erbB-4
Clone HFR-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS637P

 
thermo scientific™ lab vision™
MS637P
200μg/mL 1mL Each for $1,200.57

MS637P0

 
thermo scientific™ lab vision™
MS637P0
200μg/mL 100µL Each for $271.02

MS637P1

 
thermo scientific™ lab vision™
MS637P1
200μg/mL 500µL Each for $765.90

MS637P1ABX

 
thermo scientific™ lab vision™
MS637P1ABX
1mg/mL 100µL Each for $843.69

MS637PABX

 
thermo scientific™ lab vision™
MS637PABX
1mg/mL 200µL Each for $1,283.88
Description & Specifications

Specifications

Antigen HER-4/c-erbB-4
Clone HFR-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline EGFR Family
Type Primary
Immunogen A synthetic peptide corresponding to aa 1249-1264 (Cys-RSTLQHPDYLQEYST) from the human c-erbB-4/HER-4 oncoprotein
Isotype IgG2b

Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ HER-4/c-erbB-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-erbB-4 is the fourth member of class I receptor kinase family (ErbB). HER-4 is most predominantly expressed in several breast carcinoma cell lines, and in normal skeletal muscle, heart, pituitary, brain, and cerebellum. Breast tumor cell lines T47-D, MDA-MB-453, BT-474 and H3396 are found to have the highest levels of mRNA, and intermediate levels are seen in MCF-7, MDA-MB-330 and MDA-MB-361. Expression of erbB-4 is low or absent in some breast tumor cell lines such as MDA-MB-231, MDA-MB-157, MDA-MB-468, and SKBR-3

Host Species: Mouse

Clone: HFR-1

Isotype: IgG2b

Species Reactivity: Human and Mouse. Others not tested.

Epitope: aa 1249-1264

Immunogen: A synthetic peptide corresponding to aa 1249-1264 (Cys-RSTLQHPDYLQEYST) from the human HER-4/c-erbB-4 oncoprotein

Molecular Weight: 185kDa

Positive Control: Breast CA or Skeletal muscle

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO