missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-4/c-erbB-4 Ab-3, Mouse Monoclonal Antibody

Recommended for Flow Cytometry, Thermo Scientific™ HER-4/c-erbB-4 Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.

$274.73 - $1,556.22

Specifications

Antigen HER-4/c-erbB-4 Ab-3
Clone H4.72.8; same as Ab72
Classification Monoclonal
Cross Reactivity Human
Format Concentrated
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
MS304B Thermo Scientific™ Lab Vision™
MS304B
200μg/mL Biotin 1mL Each for $1,556.22
MS304B0 Thermo Scientific™ Lab Vision™
MS304B0
200μg/mL Biotin 100μL Each for $371.39
MS304B1 Thermo Scientific™ Lab Vision™
MS304B1
200μg/mL Biotin 500μL Each for $1,022.55
MS304P Thermo Scientific™ Lab Vision™
MS304P
200μg/mL Unlabeled 1mL Each for $1,220.65
MS304P0 Thermo Scientific™ Lab Vision™
MS304P0
200μg/mL Unlabeled 100μL Each for $274.73
MS304P1 Thermo Scientific™ Lab Vision™
MS304P1
200μg/mL Unlabeled 500μL Each for $777.37
MS304P1ABX Thermo Scientific™ Lab Vision™
MS304P1ABX
1mg/mL Unlabeled 100μL Each for $857.35
MS304PABX Thermo Scientific™ Lab Vision™
MS304PABX
1mg/mL Unlabeled 200μL Each for $1,300.96
Specifications

Description

Recommended for Flow Cytometry, Thermo Scientific™ HER-4/c-erbB-4 Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-erbB-4 is the fourth member of class I receptor kinase family (ErbB). HER-4 is most predominantly expressed in several breast carcinoma cell lines, and in normal skeletal muscle, heart, pituitary, brain, and cerebellum. Breast tumor cell lines T47-D, MDA-MB-453, BT-474 and H3396 are found to have the highest levels of mRNA, and intermediate levels are seen in MCF-7, MDA-MB-330 and MDA-MB-361. Expression of erbB-4 is low or absent in some breast tumor cell lines such as MDA-MB-231, MDA-MB-157, MDA-MB-468, and SKBR-3

Host Species: Mouse

Clone: H4.72.8; same as Ab72

Isotype: IgG2a

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Extracellular domain of recombinant human HER-4/c-erbB-4 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D or MDA-MB-453 cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Western Blotting (Not suitable)
  • Flow Cytometry
  • Inhibits Heregulin (NDF) Binding

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

HER-4/c-erbB-4 Ab-3
Monoclonal
Concentrated
RUO
Primary
IgG2a
H4.72.8; same as Ab72
Human
Murine
EGFR Family
Extracellular domain of recombinant human c-erbB-4/HER-4 oncoprotein