Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-4/c-erbB-4 Ab-3, Mouse Monoclonal Antibody


Recommended for Flow Cytometry, Thermo Scientific™ HER-4/c-erbB-4 Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  MS304P1ABX

 View more versions of this product

Catalog No. MS304P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Immunogen Extracellular domain of recombinant human c-erbB-4/HER-4 oncoprotein
Antigen HER-4/c-erbB-4 Ab-3
Clone H4.72.8; same as Ab72
Quantity 100µL
Regulatory Status RUO
Isotype IgG2a
Format Concentrated
Host Species Mouse
Classification Monoclonal
Concentration 1mg/mL
Research Discipline EGFR Family
Type Primary
Conjugate Unlabeled

Recommended for Flow Cytometry, Thermo Scientific™ HER-4/c-erbB-4 Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-erbB-4 is the fourth member of class I receptor kinase family (ErbB). HER-4 is most predominantly expressed in several breast carcinoma cell lines, and in normal skeletal muscle, heart, pituitary, brain, and cerebellum. Breast tumor cell lines T47-D, MDA-MB-453, BT-474 and H3396 are found to have the highest levels of mRNA, and intermediate levels are seen in MCF-7, MDA-MB-330 and MDA-MB-361. Expression of erbB-4 is low or absent in some breast tumor cell lines such as MDA-MB-231, MDA-MB-157, MDA-MB-468, and SKBR-3

Host Species: Mouse

Clone: H4.72.8; same as Ab72

Isotype: IgG2a

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Extracellular domain of recombinant human HER-4/c-erbB-4 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D or MDA-MB-453 cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Western Blotting (Not suitable)
  • Flow Cytometry
  • Inhibits Heregulin (NDF) Binding

Note:

USA: RUO; Int'l: RUO