Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-4/c-erbB-4 Ab-2, Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ HER-4/c-erbB-4 Ab-2, Rabbit Polyclonal Antibody.

$276.86 - $1,226.49

Specifications

Antigen HER-4/c-erbB-4 Ab-2
Clone Polyclonal
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB284P

 
thermo scientific™ lab vision™
RB284P
Purified Ab with BSA and Azide 1mL Each for $1,153.10

RB284P0

 
thermo scientific™ lab vision™
RB284P0
Purified Ab with BSA and Azide 100µL Each for $276.86

RB284P1

 
thermo scientific™ lab vision™
RB284P1
Purified Ab with BSA and Azide 500µL Each for $767.94

RB284P1ABX

 
thermo scientific™ lab vision™
RB284P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $861.92

RB284PABX

 
thermo scientific™ lab vision™
RB284PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,226.49
Description & Specifications

Specifications

Antigen HER-4/c-erbB-4 Ab-2
Clone Polyclonal
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Host Species Rabbit
Type Primary
Immunogen A synthetic peptide (aa 1285-1308) from the C-terminus of human c-erbB-4/HER-4 oncoprotein
Isotype IgG

Provide accurate, reproducible results with the Thermo Scientific™ HER-4/c-erbB-4 Ab-2, Rabbit Polyclonal Antibody.

c-erbB-4 is the fourth member of class I receptor kinase family (ErbB). HER-4 is most predominantly expressed in several breast carcinoma cell lines, and in normal skeletal muscle, heart, pituitary, brain, and cerebellum. Breast tumor cell lines T47-D, MDA-MB-453, BT-474 and H3396 are found to have the highest levels of mRNA, and intermediate levels are seen in MCF-7, MDA-MB-330 and MDA-MB-361. Expression of erbB-4 is low or absent in some breast tumor cell lines such as MDA-MB-231, MDA-MB-157, MDA-MB-468, and SKBR-3

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Rat and Mouse. Others not tested.

Epitope: aa 1285-1308

Immunogen: A synthetic peptide (aa 1285-1308) from the C-terminus of human HER-4/c-erbB-4 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D or MDA-MB-453 cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Western Blotting (Not verified)

Note:

USA: RUO; Int'l: RUO