Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-4/c-erbB-4 Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Mouse Monoclonal Antibody with the Thermo Scientific™ HER-4/c-erbB-4 Ab-1, Mouse Monoclonal Antibody.

$277.90 - $1,544.20

Specifications

Antigen HER-4/c-erbB-4 Ab-1
Clone H4.77.16 (Ab77)
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS270B

 
thermo scientific™ lab vision™
MS270B
200μg/mL Biotin 1mL Each for $1,544.20

MS270B0

 
thermo scientific™ lab vision™
MS270B0
200μg/mL Biotin 100µL Each for $368.51

MS270B1

 
thermo scientific™ lab vision™
MS270B1
200μg/mL Biotin 500µL Each for $1,091.63

MS270P

 
thermo scientific™ lab vision™
MS270P
200μg/mL Unlabeled 1mL Each for $1,228.02

MS270P0

 
thermo scientific™ lab vision™
MS270P0
200μg/mL Unlabeled 100µL Each for $277.90

MS270P1

 
thermo scientific™ lab vision™
MS270P1
200μg/mL Unlabeled 500µL Each for $777.47

MS270P1ABX

 
thermo scientific™ lab vision™
MS270P1ABX
1mg/mL Unlabeled 100µL Each for $852.08

MS270PABX

 
thermo scientific™ lab vision™
MS270PABX
1mg/mL Unlabeled 200µL Each for $1,289.61
Description & Specifications

Specifications

Antigen HER-4/c-erbB-4 Ab-1
Clone H4.77.16 (Ab77)
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Flow Cytometry
Applications Immunoprecipitation
Classification Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Immunogen Extracellular fragment of recombinant human c-erbB-4/HER-4 oncoprotein
Isotype IgG1

Provide accurate, reproducible results with the Mouse Monoclonal Antibody with the Thermo Scientific™ HER-4/c-erbB-4 Ab-1, Mouse Monoclonal Antibody.

c-erbB-4 is the fourth member of class I receptor kinase family (ErbB). Down regulation of ErbB-4 in estrogen receptor-positive cancer cells inhibits proliferation.

Host Species: Mouse

Clone: H4.77.16; same as Ab77

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Extracellular domain

Immunogen: Extracellular fragment of recombinant human HER-4/c-erbB-4 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D or MDA-MB-453 cells

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Western Blotting (Not suitable)
  • Immunoprecipitation
  • Flow Cytometry
  • Accelerates Internalization of HER-1
  • Stimulates Tyrosine Kinase Activity

Note:

USA: RUO; Int'l: RUO