missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-4/c-erbB-4 Ab-1 Mouse Monoclonal Antibody, 200µg/mL, BSA and azide

Thermo Scientific™ HER-4/c-erbB-4 Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Supplier:  Thermo Scientific™ MS270P1

Catalog No. MS270P1



Description

Description

Thermo Scientific™ HER-4/c-erbB-4 Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results. Mouse Monoclonal Antibody

c-erbB-4 is the fourth member of class I receptor kinase family (ErbB). Down regulation of ErbB-4 in estrogen receptor-positive cancer cells inhibits proliferation.

Host Species: Mouse

Clone: H4.77.16; same as Ab77

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Extracellular domain

Immunogen: Extracellular fragment of recombinant human HER-4/c-erbB-4 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D or MDA-MB-453 cells

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Western Blotting (Not suitable)
  • Immunoprecipitation
  • Flow Cytometry
  • Accelerates Internalization of HER-1
  • Stimulates Tyrosine Kinase Activity

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

HER-4/c-erbB-4 Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Primary
Concentrated
Flow Cytometry, Immunoprecipitation
H4.77.16 (Ab77)
Unconjugated
Extracellular fragment of recombinant human c-erbB-4/HER-4 oncoprotein
RUO
Human, Mouse, Rat
IgG1
SDS
Documents

Documents

Promotions

Promotions