Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-3/c-erbB-3 Ab-7, Mouse Monoclonal Antibody

Recommended for Western Blotting and Immunoprecipitation (Denatured verified), Thermo Scientific™ HER-3/c-erbB-3 Ab-7, Mouse Monoclonal Antibody provides accurate, reproducible results.

$265.00 - $1,230.90

Specifications

Antigen HER-3/c-erbB-3 Ab-7
Clone 2C3
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS313P

 
thermo scientific™ lab vision™
MS313P
200μg/mL 1mL Each for $1,163.16

MS313P0

 
thermo scientific™ lab vision™
MS313P0
200μg/mL 100µL Each for $265.00

MS313P1

 
thermo scientific™ lab vision™
MS313P1
200μg/mL 500µL Each for $740.90

MS313P1ABX

 
thermo scientific™ lab vision™
MS313P1ABX
1mg/mL 100µL Each for $809.24

MS313PABX

 
thermo scientific™ lab vision™
MS313PABX
1mg/mL 200µL Each for $1,230.90
Description & Specifications

Specifications

Antigen HER-3/c-erbB-3 Ab-7
Clone 2C3
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Immunogen Recombinant rat c-erbB-3/HER-3 oncoprotein
Isotype IgG1

Recommended for Western Blotting and Immunoprecipitation (Denatured verified), Thermo Scientific™ HER-3/c-erbB-3 Ab-7, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-erbB-3/HER-3 is the third member of the type I family of growth factor receptors. It binds to ligands in the heregulin family. c-erbB-3 is over-expressed in a variety of tumors including breast, stomach, pancreas, and colon. Heregulin & EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents.

Host Species: Mouse

Clone: 2C3

Isotype: IgG1

Species Reactivity: Human, Rat and Mouse. Others not tested.

Epitope: aa 1234-1259

Immunogen: Recombinant rat HER-3/c-erbB-3 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D, SKBR3 or MAD109 cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO