Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-3/c-erbB-3 Ab-7, Mouse Monoclonal Antibody


Recommended for Western Blotting and Immunoprecipitation (Denatured verified), Thermo Scientific™ HER-3/c-erbB-3 Ab-7, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  MS313P1ABX

 View more versions of this product

Catalog No. MS313P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Isotype IgG1
Applications Immunoprecipitation
Applications Western Blotting
Clone 2C3
Antigen HER-3/c-erbB-3 Ab-7
Host Species Mouse
Quantity 100µL
Format Concentrated
Immunogen Recombinant rat c-erbB-3/HER-3 oncoprotein
Conjugate Unlabeled
Type Primary
Concentration 1mg/mL
Research Discipline Oncogenesis
Classification Monoclonal

Recommended for Western Blotting and Immunoprecipitation (Denatured verified), Thermo Scientific™ HER-3/c-erbB-3 Ab-7, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-erbB-3/HER-3 is the third member of the type I family of growth factor receptors. It binds to ligands in the heregulin family. c-erbB-3 is over-expressed in a variety of tumors including breast, stomach, pancreas, and colon. Heregulin & EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents.

Host Species: Mouse

Clone: 2C3

Isotype: IgG1

Species Reactivity: Human, Rat and Mouse. Others not tested.

Epitope: aa 1234-1259

Immunogen: Recombinant rat HER-3/c-erbB-3 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D, SKBR3 or MAD109 cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO