Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-3/c-erbB-3 Ab-5, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ HER-3/c-erbB-3 Ab-5, Mouse Monoclonal Antibody.

$262.51 - $1,263.76

Specifications

Antigen HER-3/c-erbB-3 Ab-5
Clone H3.105.5; same as Ab105
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Immunofluorescence
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS303P

 
thermo scientific™ lab vision™
MS303P
200μg/mL 1mL Each for $1,178.27

MS303P0

 
thermo scientific™ lab vision™
MS303P0
200μg/mL 100μL Each for $262.51

MS303P1

 
thermo scientific™ lab vision™
MS303P1
200μg/mL 500μL Each for $745.82

MS303P1ABX

 
thermo scientific™ lab vision™
MS303P1ABX
1mg/mL 100μL Each for $809.73

MS303PABX

 
thermo scientific™ lab vision™
MS303PABX
1mg/mL 200μL Each for $1,263.76
Description & Specifications

Specifications

Antigen HER-3/c-erbB-3 Ab-5
Clone H3.105.5; same as Ab105
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Immunofluorescence
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline EGFR Family
Type Primary
Immunogen Extracellular domain of recombinant human c-erbB-3/HER-3 oncoprotein
Isotype IgG1

Provide accurate, reproducible results with the Thermo Scientific™ HER-3/c-erbB-3 Ab-5, Mouse Monoclonal Antibody.

c-erbB-3/HER-3 is the third member of the type I family of growth factor receptors. It binds to ligands in the heregulin family. c-erbB-3 is over-expressed in a variety of tumors including breast, stomach, pancreas, and colon. Heregulin & EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents.

Host Species: Mouse

Clone: H3.105.5; same as Ab105

Isotype: IgG1

Species Reactivity: Human and Mouse. Others not tested.

Epitope: Extracellular domain

Immunogen: Extracellular domain of recombinant human HER-3/c-erbB-3 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • Inhibits Heregulin (NDF) Binding
  • Effect on Tumor Growth (Noninhibitory)

Note:

USA: RUO; Int'l: RUO