missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-3/c-erbB-3 Ab-5, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ HER-3/c-erbB-3 Ab-5, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS303P1ABX

 View more versions of this product

Catalog No. MS303P1ABX


 

Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ HER-3/c-erbB-3 Ab-5, Mouse Monoclonal Antibody.

c-erbB-3/HER-3 is the third member of the type I family of growth factor receptors. It binds to ligands in the heregulin family. c-erbB-3 is over-expressed in a variety of tumors including breast, stomach, pancreas, and colon. Heregulin & EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents.

Host Species: Mouse

Clone: H3.105.5; same as Ab105

Isotype: IgG1

Species Reactivity: Human and Mouse. Others not tested.

Epitope: Extracellular domain

Immunogen: Extracellular domain of recombinant human HER-3/c-erbB-3 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • Inhibits Heregulin (NDF) Binding
  • Effect on Tumor Growth (Noninhibitory)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

HER-3/c-erbB-3 Ab-5
Monoclonal
1mg/mL
Concentrated
Extracellular domain of recombinant human c-erbB-3/HER-3 oncoprotein
100μL
EGFR Family
Monoclonal
Immunofluorescence,Immunoprecipitation
H3.105.5; same as Ab105
Unlabeled
Murine
IgG1
RUO
Primary
Human,Murine