Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-3/c-erbB-3 Ab-4, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ HER-3/c-erbB-3 Ab-4, Mouse Monoclonal Antibody.

$268.22 - $1,504.59

Specifications

Antigen HER-3/c-erbB-3 Ab-4
Clone H3.90.6; same as Ab90
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS262B

 
thermo scientific™ lab vision™
MS262B
200μg/mL Biotin 1mL Each for $1,504.59

MS262B0

 
thermo scientific™ lab vision™
MS262B0
200μg/mL Biotin 100μL Each for $358.78

MS262B1

 
thermo scientific™ lab vision™
MS262B1
200μg/mL Biotin 500μL Each for $977.06

MS262P

 
thermo scientific™ lab vision™
MS262P
200μg/mL Unlabeled 1mL Each for $1,185.69

MS262P0

 
thermo scientific™ lab vision™
MS262P0
200μg/mL Unlabeled 100μL Each for $268.22

MS262P1

 
thermo scientific™ lab vision™
MS262P1
200μg/mL Unlabeled 500μL Each for $754.35

MS262P1ABX

 
thermo scientific™ lab vision™
MS262P1ABX
1mg/mL Unlabeled 100μL Each for $830.22

MS262PABX

 
thermo scientific™ lab vision™
MS262PABX
1mg/mL Unlabeled 200μL Each for $1,260.10
Description & Specifications

Specifications

Antigen HER-3/c-erbB-3 Ab-4
Clone H3.90.6; same as Ab90
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
Classification Monoclonal
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Immunogen Extracellular fragment of recombinant human c-erbB-3/HER-3 oncoprotein
Isotype IgG2a

Provide accurate, reproducible results with the Thermo Scientific™ HER-3/c-erbB-3 Ab-4, Mouse Monoclonal Antibody.

c-erbB-3/HER-3 is the third member of the type I family of growth factor receptors. It binds to ligands in the heregulin family. c-erbB-3 is over-expressed in a variety of tumors including breast, stomach, pancreas, and colon. Heregulin & EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents.

Host Species: Mouse

Clone: H3.90.6; same as Ab90

Isotype: IgG2a

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Extracellular fragment of recombinant human HER-3/c-erbB-3 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • Effect on Heregulin (NDF) Binding (Noninhibitory)
  • Effect on Tumor Growth (Noninhibitory)

Note:

USA: RUO; Int'l: RUO