Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-3/c-erbB-3 Ab-2, Mouse Monoclonal Antibody

Recommended for Western Blotting and Immunoprecipitation (Native and denatured), Thermo Scientific™ HER-3/c-erbB-3 Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

$257.18 - $1,168.29

Specifications

Antigen HER-3/c-erbB-3 Ab-2
Clone 2F12
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS201P

 
thermo scientific™ lab vision™
MS201P
200μg/mL 1mL Each for $1,097.06

MS201P0

 
thermo scientific™ lab vision™
MS201P0
200μg/mL 100µL Each for $257.18

MS201P1

 
thermo scientific™ lab vision™
MS201P1
200μg/mL 500µL Each for $715.16

MS201P1ABX

 
thermo scientific™ lab vision™
MS201P1ABX
1mg/mL 100µL Each for $778.14

MS201PABX

 
thermo scientific™ lab vision™
MS201PABX
1mg/mL 200µL Each for $1,168.29
Description & Specifications

Specifications

Antigen HER-3/c-erbB-3 Ab-2
Clone 2F12
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Immunogen Recombinant rat c-erbB-3/HER-3 oncoprotein
Isotype IgG2a kappa

Recommended for Western Blotting and Immunoprecipitation (Native and denatured), Thermo Scientific™ HER-3/c-erbB-3 Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-erbB-3/HER-3 is the third member of the type I family of growth factor receptors. It binds to ligands in the heregulin family. c-erbB-3 is over-expressed in a variety of tumors including breast, stomach, pancreas, and colon. Heregulin & EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents.

Host Species: Mouse

Clone: 2F12

Isotype: IgG2a/κ

Species Reactivity: Human, Cow, Rat and Mouse. Others not tested.

Epitope: aa 1295-1323

Immunogen: Recombinant rat HER-3/c-erbB-3 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO