Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-2/c-erbB-2/neu5 Ab-15, Mouse Monoclonal Antibody

Recommended for Immunohistochemistry, Western Blotting and Immunoprecipitation (Native and Denatured), Thermo Scientific™ HER-2/c-erbB-2/neu5 Ab-15, Mouse Monoclonal Antibody provides accurate, reproducible results. 

$271.53 - $1,539.35

Specifications

Antigen HER-2/c-erbB-2/neu5 Ab-15
Clone 3B5
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS599B

 
thermo scientific™ lab vision™
MS599B
Biotinylated Ab with BSA and Azide 1mL Each for $1,539.35

MS599B0

 
thermo scientific™ lab vision™
MS599B0
Biotinylated Ab with BSA and Azide 100μL Each for $363.39

MS599B1

 
thermo scientific™ lab vision™
MS599B1
Biotinylated Ab with BSA and Azide 500μL Each for $1,000.47

MS599P

 
thermo scientific™ lab vision™
MS599P
Purified Ab with BSA and Azide 1mL Each for $1,204.51

MS599P0

 
thermo scientific™ lab vision™
MS599P0
Purified Ab with BSA and Azide 100μL Each for $271.53

MS599P1

 
thermo scientific™ lab vision™
MS599P1
Purified Ab with BSA and Azide 500μL Each for $763.94

MS599P1ABX

 
thermo scientific™ lab vision™
MS599P1ABX
Ab Without BSA/Azide (Frozen) 100μL Each for $840.16

MS599PABX

 
thermo scientific™ lab vision™
MS599PABX
Ab Without BSA/Azide (Frozen) 200μL Each for $1,270.53
Description & Specifications

Specifications

Antigen HER-2/c-erbB-2/neu5 Ab-15
Clone 3B5
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Rat
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline EGFR Family
Type Primary
Immunogen A synthetic peptide from the C-terminus of human c-erbB-2 protein. This Sequence Is Identical In Rat Neu Protein.
Isotype IgG1

Recommended for Immunohistochemistry, Western Blotting and Immunoprecipitation (Native and Denatured), Thermo Scientific™ HER-2/c-erbB-2/neu5 Ab-15, Mouse Monoclonal Antibody provides accurate, reproducible results. 

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Mouse

Clone: 3B5

Isotype: IgG1

Species Reactivity: Human, Monkey, Mouse and Rat. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide from the C-terminus of human HER-2 protein. This sequence is identical in rat neu protein.

Molecular Weight: 185 kDa

Positive Control: SKBR-3 or MAD109 cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunoprecipitation (Native and Denatured)

Note:

USA: RUO; Int'l: RUO