Learn More
HER-2/c-erbB-2/neu5 Ab-15 Mouse Monoclonal Antibody, Epredia™
Recommended for Immunohistochemistry, Western Blotting and Immunoprecipitation (Native and Denatured), Epredia™ HER-2/c-erbB-2/neu5 Ab-15, Mouse Monoclonal Antibody provides accurate, reproducible results.
Supplier: Thermo Fisher Scientific MS599P1ABX
Description
c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the proteinHost Species: Mouse
Clone: 3B5
Isotype: IgG1
Species Reactivity: Human, Monkey, Mouse and Rat. Others not tested.
Epitope: C-terminus
Immunogen: A synthetic peptide from the C-terminus of human HER-2 protein. This sequence is identical in rat neu protein.
Molecular Weight: 185 kDa
Positive Control: SKBR-3 or MAD109 cells, or Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry
- Western Blotting
- Immunoprecipitation (Native and Denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
HER-2/c-erbB-2/neu5 Ab-15 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
EGFR Family | |
Human, Mouse, Non-human Primate, Rat | |
IgG1 |
Immunoprecipitation, Western Blot | |
3B5 | |
Unconjugated | |
A synthetic peptide from the C-terminus of human c-erbB-2 protein. This Sequence Is Identical In Rat Neu Protein. | |
RUO | |
Primary | |
Concentrated |