Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-2/c-erbB-2/neu, Rabbit Polyclonal Antibody

Recommended for Western Blotting and Immunoprecipitation, Thermo Scientific™ HER-2/c-erbB-2/neu, Rabbit Polyclonal Antibody provides accurate, reproducible results.

$271.27 - $1,261.51

Specifications

Antigen HER-2/c-erbB-2/neu
Concentration 200µg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

RB9040P

 
thermo scientific™ lab vision™
RB9040P
1mL Each for $1,261.51

RB9040P0

 
thermo scientific™ lab vision™
RB9040P0
100µL Each for $271.27

RB9040P1

 
thermo scientific™ lab vision™
RB9040P1
500µL Each for $819.45
Description & Specifications

Specifications

Antigen HER-2/c-erbB-2/neu
Concentration 200µg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Immunogen A synthetic peptide from C-terminus of human HER-2 protein. This sequence is identical in rat neu protein.
Isotype IgG

Recommended for Western Blotting and Immunoprecipitation, Thermo Scientific™ HER-2/c-erbB-2/neu, Rabbit Polyclonal Antibody provides accurate, reproducible results.

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. It is closely related instructure to the epidermal growth factor receptor. c-erbB-2 oncoprotein is detectable in a proportion of breast and other adenocarconomas, as well as transitional cell carcinomas. In the case of breast cancer, expression determined by immunohistochemistry has been shown to be associated with poor prognosis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Monkey, Rat and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human HER-2 protein. This sequence is identical in rat neu protein.

Molecular Weight: 185 kDa

Positive Control: SKBR3 cells

Recommended for:

  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO