Learn More
Epredia™ Lab Vision™ HER-2/c-erbB-2/neu (Phospho-specific) Ab-18 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Provide accurate, reproducible results with the Epredia™ HER-2/c-erbB-2/neu (Phospho-specific) Ab-18, Mouse Monoclonal Antibody.
Supplier: Epredia™ MS1072B1
Description
c-erbB-2 (HER-2/neu) is a 185 kDa receptor tyrosine kinase belonging to the epidermal growth factor receptor family, which also includes EGFR (HER-1), c-erbB-3 (HER-3), and c-erbB-4 (HER-4). Amplification of the HER-2 gene with overexpression of the protein occurs in 20-30% of breast cancers, and tumor progression in mammary cancer is associated with elevated levels of tyrosine-phosphorylated neu and erbB-3. Tyr1248 is the phosphorylation site that is most tightly linked to oncogenic transformation and coupling to the ras/MAP kinase signaling pathway. Cases of invasive breast carcinoma with the phosphorylated receptor have displayed aggressive clinicopathological features and adverse prognoses. Activation of HER-2 is frequent in ductal carcinoma in situ suggesting that HER-2 signaling plays a critical role in the early stages of breast tumorigenesis.Host Species: Mouse
Clone: PN2A
Isotype: IgG1
Species Reactivity: Human and Rat. Others not tested.
Epitope: aa 1242-1255 with pTyr
Immunogen: Tyrosine-phosphorylated synthetic peptide corresponding to aa 1242-1255 [Lys-TAENPE-pY-LGLDVPV] from the C-terminus of human HER-2 protein (P-Tyr1248)
Molecular Weight: 185 kDa
Positive Control: EGF-treated SKBR-3 cells. About 5% of breast CA are positive for phospho-HER-2/c-erbB-2/neu oncoprotein.
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Not verified)
- Flow Cytometry (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
HER-2/c-erbB-2/neu (Phospho-specific) Ab-18 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
EGFR Family | |
Human, Rat | |
IgG1 |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
PN2A | |
Biotin | |
Tyrosine-phosphorylated synthetic peptide corresponding to aa1242-1255 [Lys-TAENPE-pY-LGLDVPV] from the C-terminus of human c-erbB-2 protein (P-Tyr1248) | |
RUO | |
Primary | |
Concentrated |