Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-2/c-erbB-2/neu Ab-8, Mouse Monoclonal Antibody

Recommended for Immunohistochemistry, Western Blotting and Immunoprecipitation (Native and Denatured), Thermo Scientific™ HER-2/c-erbB-2/neu Ab-8, Mouse Monoclonal Antibody provides accurate, reproducible results.

$256.57 - $1,437.20

Specifications

Antigen HER-2/c-erbB-2/neu Ab-8
Clone e2-4001
Applications Immunoprecipitation
Classification Monoclonal
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS325B

 
thermo scientific™ lab vision™
MS325B
Biotinylated Ab with BSA and Azide 1mL Each for $1,437.20

MS325B0

 
thermo scientific™ lab vision™
MS325B0
Biotinylated Ab with BSA and Azide 100μL Each for $346.41

MS325B1

 
thermo scientific™ lab vision™
MS325B1
Biotinylated Ab with BSA and Azide 500μL Each for $943.43

MS325P

 
thermo scientific™ lab vision™
MS325P
Purified Ab with BSA and Azide 1mL Each for $1,139.31

MS325P0

 
thermo scientific™ lab vision™
MS325P0
Purified Ab with BSA and Azide 100μL Each for $256.57

MS325P1

 
thermo scientific™ lab vision™
MS325P1
Purified Ab with BSA and Azide 500μL Each for $721.33

MS325P1ABX

 
thermo scientific™ lab vision™
MS325P1ABX
Ab Without BSA/Azide (Frozen) 100μL Each for $784.67

MS325PABX

 
thermo scientific™ lab vision™
MS325PABX
Ab Without BSA/Azide (Frozen) 200μL Each for $1,201.94
Description & Specifications

Specifications

Antigen HER-2/c-erbB-2/neu Ab-8
Clone e2-4001
Applications Immunoprecipitation
Classification Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Immunogen Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein
Isotype IgG1

Recommended for Immunohistochemistry, Western Blotting and Immunoprecipitation (Native and Denatured), Thermo Scientific™ HER-2/c-erbB-2/neu Ab-8, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-erbB-2 is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein. The c-erbB-2 protein is overexpressed in a variety of carcinomas especially those of breast and ovary.

Host Species: Mouse

Clone: e2-4001

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Cytoplasmic domain

Immunogen: Cytoplasmic domain of recombinant human HER-2/c-erbB-2 oncoprotein

Molecular Weight: 185 kDa

Positive Control: SKBR-3 cells, or T47D cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunoprecipitation (Native and Denatured)

Note:

USA: RUO; Int'l: RUO