missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-2/c-erbB-2/neu Ab-6, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ HER-2/c-erbB-2/neu Ab-6, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS307P1ABX

 View more versions of this product

Catalog No. MS307P1ABX


 

Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ HER-2/c-erbB-2/neu Ab-6, Mouse Monoclonal Antibody.

c-erbB-2, second member (c-erbB-2/HER-2/neu) of the c-erbB family is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein.

Host Species: Mouse

Clone: N28

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not tested.

Epitope: Extracellular domain

Immunogen: Intact SKBR-3 breast cancer cells

Molecular Weight: 185 kDa

Positive Control: SKBR-3 cells

Cellular Localization: Membrane

Recommended for:

  • Immunoprecipitation (Native and denatured)
  • Stimulates (10-fold) Tyrosine Kinase Activity
  • Effect on Tumor Growth (∽40% stimulatory)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

HER-2/c-erbB-2/neu Ab-6
Monoclonal
1mg/mL
Concentrated
Intact SKBR-3 breast cancer cells
100μL
Oncogenesis
Monoclonal
Immunoprecipitation
N28
Unlabeled
Murine
IgG1
RUO
Primary
Human