missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-2/c-erbB-2/neu Ab-5, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ HER-2/c-erbB-2/neu Ab-5, Mouse Monoclonal Antibody.

$274.29 - $1,269.14

Specifications

Antigen HER-2/c-erbB-2/neu Ab-5
Clone N24
Applications Immunofluorescence,Immunoprecipitation,Immunofluorescence,Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS302P Thermo Scientific™ Lab Vision™
MS302P
200μg/mL 1mL Each for $1,206.57
MS302P0 Thermo Scientific™ Lab Vision™
MS302P0
200μg/mL 100μL Each for $274.29
MS302P1 Thermo Scientific™ Lab Vision™
MS302P1
200μg/mL 500μL Each for $769.24
MS302P1ABX Thermo Scientific™ Lab Vision™
MS302P1ABX
1mg/mL 100μL Each for $863.43
MS302PABX Thermo Scientific™ Lab Vision™
MS302PABX
1mg/mL 200μL Each for $1,269.14
Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ HER-2/c-erbB-2/neu Ab-5, Mouse Monoclonal Antibody.

c-erbB-2 is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Mouse

Clone: N24

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not tested.

Epitope: Extracellular domain

Immunogen: Intact SKBR-3 breast cancer cells

Molecular Weight: 185 kDa

Positive Control: SKBR-3 cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence
  • Immunoprecipitation
  • Accelerates (∽50%) HER-2 Internalization
  • Inhibits (∽80%) Tumorigenic Cell Growth

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

HER-2/c-erbB-2/neu Ab-5
Immunofluorescence,Immunoprecipitation,Immunofluorescence,Immunoprecipitation
Unlabeled
Murine
EGFR Family
Intact SKBR-3 breast cancer cells
N24
Monoclonal
Concentrated
RUO
Primary
IgG1