Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-2/c-erbB-2/neu Ab-20, Mouse Monoclonal Antibody

Recommended for Western Blotting (Extracellular), Thermo Scientific™ HER-2/c-erbB-2/neu Ab-20, Mouse Monoclonal Antibody provides accurate, reproducible results.

$276.12 - $1,473.39

Specifications

Antigen HER-2/c-erbB-2/neu Ab-20
Clone L87 + 2ERB19
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1350P

 
thermo scientific™ lab vision™
MS1350P
200μg/mL 1mL Each for $1,406.16

MS1350P0

 
thermo scientific™ lab vision™
MS1350P0
200μg/mL 100µL Each for $276.12

MS1350P1

 
thermo scientific™ lab vision™
MS1350P1
200μg/mL 500µL Each for $869.57

MS1350P1ABX

 
thermo scientific™ lab vision™
MS1350P1ABX
1mg/mL 100µL Each for $971.59

MS1350PABX

 
thermo scientific™ lab vision™
MS1350PABX
1mg/mL 200µL Each for $1,473.39
Description & Specifications

Specifications

Antigen HER-2/c-erbB-2/neu Ab-20
Clone L87 + 2ERB19
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Immunogen Extracellular domain of recombinant human c-erbB-2 oncoprotein

Recommended for Western Blotting (Extracellular), Thermo Scientific™ HER-2/c-erbB-2/neu Ab-20, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein.

Host Species: Mouse

Clone: L87 + 2ERB19

Isotype: IgG1 (L87); IgG1 (2ERB19)

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Extracellular domain of recombinant human HER-2 oncoprotein

Molecular Weight: 185kDa

Positive Control: SKBR-3 cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting (Extracellular)

Note:

USA: RUO; Int'l: RUO