Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-2/c-erbB-2/neu Ab-20, Mouse Monoclonal Antibody


Recommended for Western Blotting (Extracellular), Thermo Scientific™ HER-2/c-erbB-2/neu Ab-20, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  MS1350PABX

 View more versions of this product

Catalog No. MS1350PABX

  • / Each
 

Description & Specifications

Specifications

Antigen HER-2/c-erbB-2/neu Ab-20
Format Concentrated
Host Species Mouse
Applications Western Blotting
Immunogen Extracellular domain of recombinant human c-erbB-2 oncoprotein
Isotype IgG1
Quantity 200µL
Regulatory Status RUO
Clone L87 + 2ERB19
Cross Reactivity Human
Conjugate Unlabeled
Research Discipline Oncogenesis
Type Primary
Classification Monoclonal
Concentration 1mg/mL

Recommended for Western Blotting (Extracellular), Thermo Scientific™ HER-2/c-erbB-2/neu Ab-20, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein.

Host Species: Mouse

Clone: L87 + 2ERB19

Isotype: IgG1 (L87); IgG1 (2ERB19)

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Extracellular domain of recombinant human HER-2 oncoprotein

Molecular Weight: 185kDa

Positive Control: SKBR-3 cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting (Extracellular)

Note:

USA: RUO; Int'l: RUO