Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-2/c-erbB-2/neu Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ HER-2/c-erbB-2/neu Ab-2, Mouse Monoclonal Antibody.

$279.80 - $1,203.49

Specifications

Antigen HER-2/c-erbB-2/neu Ab-2
Clone 9G6.10
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS229P

 
thermo scientific™ lab vision™
MS229P
200μg/mL 1mL Each for $1,125.65

MS229P0

 
thermo scientific™ lab vision™
MS229P0
200μg/mL 100µL Each for $279.80

MS229P1

 
thermo scientific™ lab vision™
MS229P1
200μg/mL 500µL Each for $715.16

MS229P1ABX

 
thermo scientific™ lab vision™
MS229P1ABX
1mg/mL 100µL Each for $798.24

MS229PABX

 
thermo scientific™ lab vision™
MS229PABX
1mg/mL 200µL Each for $1,203.49
Description & Specifications

Specifications

Antigen HER-2/c-erbB-2/neu Ab-2
Clone 9G6.10
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Immunogen RAC311 cells transfected with SV40-neu construct, expressing human neu protein
Isotype IgG1

Provides accurate, reproducible results with the Thermo Scientific™ HER-2/c-erbB-2/neu Ab-2, Mouse Monoclonal Antibody.

c-erbB-2, second member (c-erbB-2/HER-2/neu) of the c-erbB family is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Mouse

Clone: 9G6.10

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not tested.

Epitope: Extracellular domain

Immunogen: RAC311 cells transfected with SV40-neu construct, expressing human neu protein

Molecular Weight: 185kDa

Positive Control: SKBR-3 cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen only) Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • Inhibits Colony Formation
  • Partially Inhibits Tyrosine Kinase Activity

Note:

USA: RUO; Int'l: RUO