Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-2/c-erbB-2/neu Ab-16, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ HER-2/c-erbB-2/neu Ab-16, Mouse Monoclonal Antibody.

$268.06 - $1,261.39

Specifications

Antigen HER-2/c-erbB-2/neu Ab-16
Clone L26
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS600P0A

 
thermo scientific™ lab vision™
MS600P0A
200μg/mL 100μL Each for $268.06

MS600P1A

 
thermo scientific™ lab vision™
MS600P1A
200μg/mL 500μL Each for $751.71

MS600P1ABX

 
thermo scientific™ lab vision™
MS600P1ABX
1mg/mL 100μL Each for $838.59

MS600PA

 
thermo scientific™ lab vision™
MS600PA
200μg/mL 1mL Each for $1,191.88

MS600PABX

 
thermo scientific™ lab vision™
MS600PABX
1mg/mL 200μL Each for $1,261.39
Description & Specifications

Specifications

Antigen HER-2/c-erbB-2/neu Ab-16
Clone L26
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline EGFR Family
Type Primary
Immunogen Extracellular domain of recombinant human c-erbB-2/HER-2 oncoprotein
Isotype IgG2a kappa

Provide accurate, reproducible results with the Thermo Scientific™ HER-2/c-erbB-2/neu Ab-16, Mouse Monoclonal Antibody.

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Mouse

Clone: L26

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Involved in the formation of heterodimers

Immunogen: Extracellular domain of recombinant human HER-2/c-erbB-2 oncoprotein

Molecular Weight: 185kDa

Positive Control: SKBR-3 cells

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Not suitable)
  • Immunoprecipitation (Native only)
  • Inhibits Binding of EGF (∽70%), NDF (∽40%)
  • Reduces Affinity of EGF (3-fold) and NDF (2-fold)

Note:

USA: RUO; Int'l: RUO